Learn More
Felix Storch Inc ACCUCOLD Controlled Rm Temp. ARG8PV-CRT 8cf GLD
Supplier: Felix Storch Inc ARG8PVCRT
ACCUCOLD ARG8PV-CRT (8cf) Controlled Room Temperature (CRT 68F TO 77F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. PRE-SET TO 72F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5C Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Glass Door: https://www.accucold.com/product/ARG8PV-CRT