Learn More
Felix Storch Inc ACCUCOLD Controlled Rm Temp. ARS1PV-CRT 15cf SLD
SDP
Supplier:  Felix Storch Inc ARS15PVCRT
Encompass_Preferred
ACCUCOLD ARS15PV-CRT (15cf) Controlled Room Temperature (CRT 68F TO 77F) cabinets are designed and purpose-built for ambient medication and vaccine storage in pharmacy and patient care areas. PRE-SET TO 72F. Equipped with Hi/Lo Temp Alarm, Remote Contacts, open door alarm, power failure alarm, Sensor failure alarm, Self closing door, Hospital grade cord & plug, Access Port, Antimicrobial Handle. +/-1.5C Variation The published temperature variation is derived from the maximum deviation of an NTC sensor in a 1 oz. vial located nearest the chamber geometric center during a 24 hour test period. LED interior Light. Solid Door: https://www.accucold.com/product/ARS15PV-CRT
Catalog No. NC2256204
Add to Cart
Request a Quote