Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Controlled Tissue Bath
SDP
Catalog No. 50949922
Encompass_Preferred
The bath for EMS7000smz and EMS5000mz Vibrating Microtome uses"Peltier" thermoelectric elements. The stainless steel tissue bath and mount are detachable to allow sterilization by autoclave if required.
Catalog No. 50-949-922 Supplier Electron Microscopy Sciences Supplier No. 7610A
The bath for EMS7000smz and EMS5000mz Vibrating Microtome uses"Peltier" thermoelectric elements. The stainless steel tissue bath and mount are detachable to allow sterilization by autoclave if required.