Learn More
Cole-Parmer Myron L 753II-11 Digital Resistivity Controller, 0 to 20 MOhm
Catalog No. 50631781 Shop All Cole-Parmer Products
Encompass_Preferred
These monitor/controllers are perfect for ultrapure water applications. Precise dual-thermistor circuit for temperature compensation matches pure water temperature coefficients-ensures accurate measurements over the entire range. Order resistivity cells separately.
Catalog No. 50-631-781 Supplier Cole-Parmer Supplier No. 1901002
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These monitor/controllers are perfect for ultrapure water applications. Precise dual-thermistor circuit for temperature compensation matches pure water temperature coefficients-ensures accurate measurements over the entire range. Order resistivity cells separately.