Learn More
Wilmad Labglass CONTROLLER FLOW SJ 35/20
Supplier:  Wilmad Labglass LG4520100
Encompass_Preferred
Flow Controller, Spherical Joint. Joint Size: [SJ] 35/20. Flow controller For use w/ LG-4530 and LG-4535 chromatography columns and LG-4525 chromatography reservoirs.
Catalog No. 50-805-827