Learn More
Enterprise Technology Solutions ControlTek Bag,Dual Deposit,Vertical,1000-Bill Cap,10"x15",100/PK,CL
SDP
Supplier:  Enterprise Technology Solutions CNK585092
Encompass_Preferred
Providing a perfect balance of security and easy use, SafeLOK VT Tamper-Evident Deposit Bags are ideal for retail applications where quick training of new employees is important. Patented security features alert to attempted theft while Press2Lok technology makes using the bag easy for your associates. Press2Lok closure is simple to press and seal. Extra pocket for checks and deposit slips lets you separate cash from checks for quicker processing by your bank and quicker funds availability for you. High-visibility void message alerts you to tampering with cold or chemicals. Special thermochromic ink on the high-performance film turns red when subjected to heat. Random, printed line graphics prevent cut-and-replace tampering. Anti-static liner does not cling to hands and clothes, speeding up processing. Large serial numbers and bar codes allow easy tracking via CCTV. Security border and micro printing prevent cutting and resealing. Each bag includes a serial number and tear-off receipt.
Catalog No. 50-008-30790