Learn More
Optimize Technologies OPTI-MAX™ Cartridge Check Valve Conversion Kit, Shimadzu LC-30
SDP
Catalog No. 502006674
Encompass_Preferred
Make the switch to OPTI-MAX™ for less than ever before with OPTI-MAX™ Conversion Kits. Each kit includes all the OPTI-MAX™ check valve housings and cartridges you need to convert your HPLC instrument. By default, each kit includes our recommended OPTI-MAX™ cartridge–the stainless steel/ ceramic OPTI-MAX™ (other cartridge options are available upon request). If you'd like to substitute an alternate material the kit price will not be affected. These kits contain all the check valves you need for your HPLC pump grouped under a single part number.
Catalog No. 50-200-6674 Supplier Optimize Technologies Supplier No. 348605140
Make the switch to OPTI-MAX™ for less than ever before with OPTI-MAX™ Conversion Kits. Each kit includes all the OPTI-MAX™ check valve housings and cartridges you need to convert your HPLC instrument. By default, each kit includes our recommended OPTI-MAX™ cartridge–the stainless steel/ ceramic OPTI-MAX™ (other cartridge options are available upon request). If you'd like to substitute an alternate material the kit price will not be affected. These kits contain all the check valves you need for your HPLC pump grouped under a single part number.