Learn More
Optimize Technologies OPTI-MAX™ Cartridge Check Valve Conversion Kit, 1/8" Ceramic Ball & Seat, SS Cartridge, Waters 100µL/Dual Head Systems
SDP
Catalog No. 502006710
Encompass_Preferred
Make the switch to OPTI-MAX™ for less than ever before with OPTI-MAX™ Conversion Kits. Each kit includes all the OPTI-MAX™ check valve housings and cartridges you need to convert your HPLC instrument. By default, each kit includes our recommended stainless steel/ceramic OPTI-MAX™ cartridge (PEEK cartridges are supplied for biocompatible systems). However, if you'd like to substitute an alternate material the kit price will not be affected. These kits contain all the check valves you need for your HPLC pump grouped under a single part number.
Catalog No. 50-200-6710 Supplier Optimize Technologies Supplier No. 395602166
Make the switch to OPTI-MAX™ for less than ever before with OPTI-MAX™ Conversion Kits. Each kit includes all the OPTI-MAX™ check valve housings and cartridges you need to convert your HPLC instrument. By default, each kit includes our recommended stainless steel/ceramic OPTI-MAX™ cartridge (PEEK cartridges are supplied for biocompatible systems). However, if you'd like to substitute an alternate material the kit price will not be affected. These kits contain all the check valves you need for your HPLC pump grouped under a single part number.