Learn More
Grainger L.B. White Portable Gas Convection Heater, 200,000 BtuH, Propane
Catalog No. 502766196
Encompass_Preferred
This portable gas convection can heater is designed for temporary or supplemental heating in construction sites, warehouses, and outdoor workspaces. Delivering 200,000 BtuH of heating capacity, this direct-fired, non-vented unit efficiently provides warm air for drying, curing, and thawing applications. It features a rugged, galvanized steel housing and a cast iron burner for durability. Operation is supported by a matchless piezo pilot ignition system and a thermocouple-controlled gas valve for enhanced safety. The heater requires a 120V AC power source and operates on liquid propane with a fuel consumption rate of 9.3 lb/hr.
  • Heating capacity of 200,000 BtuH with 850 cfm airflow
  • Matchless piezo pilot ignition system
  • Heavy-gauge steel body with galvanized finish
  • Includes 10 ft. gas hose and regulator
  • CSA certified for safe operation
  • Compatible with 100 lb. propane cylinders
Catalog No. 50-276-6196 Supplier Grainger Supplier No. 16W295
This portable gas convection can heater is designed for temporary or supplemental heating in construction sites, warehouses, and outdoor workspaces. Delivering 200,000 BtuH of heating capacity, this direct-fired, non-vented unit efficiently provides warm air for drying, curing, and thawing applications. It features a rugged, galvanized steel housing and a cast iron burner for durability. Operation is supported by a matchless piezo pilot ignition system and a thermocouple-controlled gas valve for enhanced safety. The heater requires a 120V AC power source and operates on liquid propane with a fuel consumption rate of 9.3 lb/hr.
  • Heating capacity of 200,000 BtuH with 850 cfm airflow
  • Matchless piezo pilot ignition system
  • Heavy-gauge steel body with galvanized finish
  • Includes 10 ft. gas hose and regulator
  • CSA certified for safe operation
  • Compatible with 100 lb. propane cylinders