Learn More
Grainger Convctn Prtble Gas Flr Heatr NG 3150sqft
Catalog No. 502765232
Encompass_Preferred
The L.B. White Portable Gas Ducted Tent and Space Heater is a portable heating solution for tents and other temporary enclosures. It features a 30,000 BTU propane heater with a ducted fan to circulate warm air. The unit has an adjustable thermostat and can heat spaces up to 600 square feet. Safety features include a tip-over switch and an oxygen depletion sensor. This portable heater requires propane fuel and is CSA approved for use in the US and Canada. With its ducting and thermostat controls, this L.B. White heater provides an efficient way to heat tents and temporary spaces.
Catalog No. 50-276-5232 Supplier Grainger Supplier No. 16W293
The L.B. White Portable Gas Ducted Tent and Space Heater is a portable heating solution for tents and other temporary enclosures. It features a 30,000 BTU propane heater with a ducted fan to circulate warm air. The unit has an adjustable thermostat and can heat spaces up to 600 square feet. Safety features include a tip-over switch and an oxygen depletion sensor. This portable heater requires propane fuel and is CSA approved for use in the US and Canada. With its ducting and thermostat controls, this L.B. White heater provides an efficient way to heat tents and temporary spaces.