Learn More
Chemglass Life Sciences Convenience Pack, 2.0mL Clear Glass 12x32mm Shell Vial, Snap Plug
SDP
Catalog No. 50231315
Encompass_Preferred
Convenience Pack, 2.0mL Clear Glass 12x32mm Shell Vial, Snap Plug. Shell vials are the most economical alternative for use in an auto sampler. They are available in clear and amber borosilicate or polypropylene for easy disposal. Feature thicker walls for safer sample handling.
Catalog No. 50-231-315 Supplier Chemglass Life Sciences Supplier No. CV70501232
Convenience Pack, 2.0mL Clear Glass 12x32mm Shell Vial, Snap Plug. Shell vials are the most economical alternative for use in an auto sampler. They are available in clear and amber borosilicate or polypropylene for easy disposal. Feature thicker walls for safer sample handling.