Learn More
Chemglass Life Sciences Convenience Pack, 0.75mL Clear Glass 8x30mm Shell Vial, Snap Plug (for Perkin Elmer)
SDP
Catalog No. 50231308
Encompass_Preferred
Convenience Pack, 0.75mL Clear Glass 8x30mm Shell Vial, Snap Plug (for Perkin Elmer). Shell vials are the most economical alternative for use in an auto sampler. They are available in clear and amber borosilicate or polypropylene for easy disposal. Feature thicker walls for safer sample handling.
Catalog No. 50-231-308 Supplier Chemglass Life Sciences Supplier No. CV70000830
Convenience Pack, 0.75mL Clear Glass 8x30mm Shell Vial, Snap Plug (for Perkin Elmer). Shell vials are the most economical alternative for use in an auto sampler. They are available in clear and amber borosilicate or polypropylene for easy disposal. Feature thicker walls for safer sample handling.