Learn More
Binder Inc Convenient water supply
SDP
Supplier:  Binder Inc 80091128
Encompass_Preferred
Convenient water supply - External, with magnetic holder, Set consisting of 10 liter water container with magnetic holder, hose adapter with practical quick coupling and water stop function, flexible positioning, enables water to be supplied to multiple CB units simultaneously via a water container, not pre-filled.
Catalog No. 80-091-128