Learn More
Fritsch Milling And Sizing Inc Conversion Kit for Grinding Large Quantities, P-14 Classic Line
SDP
Catalog No. 501533943
Encompass
Conversion kit for grinding large quantities for PULVERISETTE 14 classic line
Consisting of pan with outlet and flange-mount nylon support sack (outside) for the paper filter bag (inside).
Catalog No. 50-153-3943 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.3510.00
Conversion kit for grinding large quantities for PULVERISETTE 14 classic line
Consisting of pan with outlet and flange-mount nylon support sack (outside) for the paper filter bag (inside).