Learn More
Integra Biosciences Corp Conversion set for 90L mm
SDP
Catalog No. 5000882050
Encompass
Conversion set for 90L mm Petri dishes for MEDIAJET vario onlyComprising the adaptation insert head plates for feeder and stacker piston and 1 tubing set
Catalog No. 50-008-82050 Supplier Integra Biosciences Corp Supplier No. 113055
Conversion set for 90L mm Petri dishes for MEDIAJET vario onlyComprising the adaptation insert head plates for feeder and stacker piston and 1 tubing set