Learn More
Med Vet International Miltex Dental Fg/Hp Converter, New Friction Type, For Handpiece, Each, 75-40
Catalog No. 502829105
Encompass_Preferred
Designed to convert friction grip shank to hand piece shank. Made from stainless steel.
Catalog No. 50-282-9105 Supplier Med Vet International Supplier No. MIL7540
Designed to convert friction grip shank to hand piece shank. Made from stainless steel.