Learn More
Hach Company Differential 1.5" pH sensor, LCP, 5-wire, 3 m cable
Catalog No. 501997460 Shop All Hach Company Products
Encompass
Differential pH sensor with patented measurement technology using 3 electrodes. Low maintanance, versatile mounting styles, great accuracy and reliability. Chemical resistant body.
Catalog No. 50-199-7460 Supplier Hach Company Supplier No. 6028P0
Differential pH sensor with patented measurement technology using 3 electrodes. Low maintanance, versatile mounting styles, great accuracy and reliability. Chemical resistant body.