Learn More
Hach Company 3/4 Inch Combination pH/ORP Sensor, Convertible Sensor Style, Ryton Body Material, General Purpose pH Glass Electode, Temperature Compensation - PT1000 ohm RTD
Catalog No. 501998062 Shop All Hach Company Products
Encompass
Ryton, Convertible Body Style, 4.5m Analog Cable, General Purpose Glass pH Electrode, Pt 1000 Temperature Compensation
Catalog No. 50-199-8062 Supplier Hach Company Supplier No. PC1R1A
Ryton, Convertible Body Style, 4.5m Analog Cable, General Purpose Glass pH Electrode, Pt 1000 Temperature Compensation