Learn More
Grainger Mimeco Convertible Hand Truck, Aluminum, 600 lb, 1,200 lb, 1 ea
Catalog No. 502792456
Encompass_Preferred
This convertible hand truck features a versatile dual design that functions as both a standard hand truck and a robust platform truck to accommodate various transport needs. Constructed from durable, powder-coated aluminum, it provides a lightweight yet strong frame for material handling tasks in industrial or commercial environments.
  • 600 lb load capacity in hand truck mode and 1,200 lb capacity as a platform truck
  • Flat back frame design with dual handles for improved control and maneuverability
  • Open straight, beveled nose plate measuring 18 in. wide by 7 1/2 in. deep
  • Platform dimensions of 46 in. long by 12 in. wide with an 11 in. platform height
  • Pneumatic polyurethane wheels for reliable performance
Catalog No. 50-279-2456 Supplier Grainger Supplier No. 1AGW2
This convertible hand truck features a versatile dual design that functions as both a standard hand truck and a robust platform truck to accommodate various transport needs. Constructed from durable, powder-coated aluminum, it provides a lightweight yet strong frame for material handling tasks in industrial or commercial environments.
  • 600 lb load capacity in hand truck mode and 1,200 lb capacity as a platform truck
  • Flat back frame design with dual handles for improved control and maneuverability
  • Open straight, beveled nose plate measuring 18 in. wide by 7 1/2 in. deep
  • Platform dimensions of 46 in. long by 12 in. wide with an 11 in. platform height
  • Pneumatic polyurethane wheels for reliable performance