Learn More
Enzo Life Sciences Convulxin (50 µg) CAS: 37206-04-5
SDP
Supplier:  Enzo Life Sciences ALX350100C050
Encompass_Preferred
Heterodimeric C-type lectin. Heterodimeric C-type lectin. Activates mammalian platelets via binding and clustering of p62/GPVI-receptors under physiological conditions. MW: ~84kDa. Formulation: Lyophilized. Source: Isolated from Crotalus durissus terrificus snake venom. Solubility: Soluble in organic buffer (HEPES). Use/Stability: Stock solutions are stable for 8 hours at +20°C, for 2 days at +4°C or for 1 month when stored at -80°C. UniProt: O93426. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -20°C
Catalog No. NC9181035