Learn More
Bioworld COOH-Coated Microplates, 10 Plate(s)
Catalog No. 50197547
Encompass
bioWorld's Carboxylated 96-well Microplates contains a COOH-coated surfaceto which the amino group present in any molecules, such as peptides or proteins, binds to through formation of amide bonds by the action of carbodiimide.Applications:Coupling molecules, having an amino group, to COOH surface
Catalog No. 50-197-547 Supplier Bioworld Supplier No. 201400161
bioWorld's Carboxylated 96-well Microplates contains a COOH-coated surfaceto which the amino group present in any molecules, such as peptides or proteins, binds to through formation of amide bonds by the action of carbodiimide.Applications:Coupling molecules, having an amino group, to COOH surface