Learn More
Biolog Inc COOKED/CHOPPED MEAT BROTH
SDP
Supplier:  Biolog Inc AS811
Encompass
This item has a minimum qty of 2 per supplier requirements.
Chopped Meat Medium(CM), Ten 7mL Tubes/PK, Holding medium for sporulation of clostridia and for testing for proteolysis of clostridia, pH: 7.0 +/- 0.3 at 25degreeC, Shelf Life: Up to one year, Stored at room temperature
Catalog No. NC9150416