Learn More
Alkali Scientific Cooked Meat Medium w/Glucose, Hemin and Vitamin K Tube, 10/Pk
SDP
Catalog No. 5000662644
Encompass_Preferred
Cooked Meat Medium with Glucose, Hemin, and Vitamin K is an enriched formulation designed for cultivating fastidious and obligate anaerobic bacteria. Each tube contains nutrient broth and sterilized beef heart tissue particles supplemented with glucose, hemin, and vitamin K1. The meat particles act as natural reducing agents that lower the oxidation reduction potential, creating an environment suitable for anaerobic growth. Glucose provides a readily available energy source that promotes rapid metabolic activity, while hemin and vitamin K serve as essential growth factors required by many anaerobic pathogens. This formulation supports the growth of demanding organisms such as Bacteroides, Prevotella, Porphyromonas, and Fusobacterium species. It is commonly used in clinical and research laboratories for primary isolation, recovery, and maintenance of anaerobic bacteria.
Catalog No. 50-006-62644 Supplier Alkali Scientific Supplier No. CMCT01
Cooked Meat Medium with Glucose, Hemin, and Vitamin K is an enriched formulation designed for cultivating fastidious and obligate anaerobic bacteria. Each tube contains nutrient broth and sterilized beef heart tissue particles supplemented with glucose, hemin, and vitamin K1. The meat particles act as natural reducing agents that lower the oxidation reduction potential, creating an environment suitable for anaerobic growth. Glucose provides a readily available energy source that promotes rapid metabolic activity, while hemin and vitamin K serve as essential growth factors required by many anaerobic pathogens. This formulation supports the growth of demanding organisms such as Bacteroides, Prevotella, Porphyromonas, and Fusobacterium species. It is commonly used in clinical and research laboratories for primary isolation, recovery, and maintenance of anaerobic bacteria.