Learn More
MSC Detco Cleaner and Degreaser, Liquid, Alkaline, 5 gal
Catalog No. 502819416
Encompass
This cleaner and degreaser is a liquid, alkaline solution designed for use in food and beverage plants. It is also biodegradable.
  • Liquid formula
  • Alkaline formula
  • No scent
  • Designed for food & beverage plants
  • Supplied in a 5 gal bucket
  • Dilutable concentrate
  • Non-acid formulation
  • VOC content 10 g/L
  • Biodegradable
Catalog No. 50-281-9416 Supplier MSC Supplier No. 33702127
This cleaner and degreaser is a liquid, alkaline solution designed for use in food and beverage plants. It is also biodegradable.
  • Liquid formula
  • Alkaline formula
  • No scent
  • Designed for food & beverage plants
  • Supplied in a 5 gal bucket
  • Dilutable concentrate
  • Non-acid formulation
  • VOC content 10 g/L
  • Biodegradable