Learn More
Chemglass Life Sciences Cool Block, Fits 96 Well PCR Plates/Tubes
SDP
Catalog No. 501532332
Encompass
Cool Block, Fits 96 Well PCR Plates/Tubes. Solid aluminum metal chamber for 96 well PCR plate or tubes. Ideal way to keep PCR tubes and plates cold during sample preparation. Place the metal chamber in an ice bucket or on Lab Armor Beads (CLS-2991). Temperature Remains Uniform Across the Block. Dimensions 6 x 3.2 x 1. Provides Stable Platform for PCR Plate or Tubes Making it Easy to Add Samples to the Wells. Can be Stored in the Refrigerator or Freezer. Can be Used on Dry Ice or Lab Armor Beads
Catalog No. 50-153-2332 Supplier Chemglass Life Sciences Supplier No. CLS362001
Cool Block, Fits 96 Well PCR Plates/Tubes. Solid aluminum metal chamber for 96 well PCR plate or tubes. Ideal way to keep PCR tubes and plates cold during sample preparation. Place the metal chamber in an ice bucket or on Lab Armor Beads (CLS-2991). Temperature Remains Uniform Across the Block. Dimensions 6 x 3.2 x 1. Provides Stable Platform for PCR Plate or Tubes Making it Easy to Add Samples to the Wells. Can be Stored in the Refrigerator or Freezer. Can be Used on Dry Ice or Lab Armor Beads