Learn More
Chemglass Life Sciences Cool Block for 96 Well Flat Bottom Plate
SDP
Catalog No. 501532330
Encompass
Cool Block for 96 Well Flat Bottom Plate. Similar to Chemglass's CLS-3620, but designed to fit a flat bottom 96 well plate. Additional holes on the sides hold 10 each 1.5 or 2mL tubes and 12 each 0.5mL tubes. Dimensions are 7.5 x 3 x 1.
Catalog No. 50-153-2330 Supplier Chemglass Life Sciences Supplier No. CLS350801
Cool Block for 96 Well Flat Bottom Plate. Similar to Chemglass's CLS-3620, but designed to fit a flat bottom 96 well plate. Additional holes on the sides hold 10 each 1.5 or 2mL tubes and 12 each 0.5mL tubes. Dimensions are 7.5 x 3 x 1.