Learn More
Electron Microscopy Sciences True North Cool Containers, 4L Pan, Artic Blue
SDP
Catalog No. 501901692
Encompass_Preferred
These rigid single mold polyurethane containers provide durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra cold and can be used with ice, LN2, ice-salt slurries, and more.
They come with a convenient lid which nests beneath bucket or pan. Minimize evaporation loss with snug fit lid and easily remove with comfortable recessed knobs. Keyed lid and base fit together to conveniently store lid under base. Use handy pour spout with melted ice, water, and solutions. Ergonomic handles safely transport bucket or pan from work area to sink. Deep internal cavity has enough room to add one of your racks. Tapered sides make stacking and storage compact.
Catalog No. 50-190-1692 Supplier Electron Microscopy Sciences Supplier No. 6201340
These rigid single mold polyurethane containers provide durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra cold and can be used with ice, LN2, ice-salt slurries, and more.
They come with a convenient lid which nests beneath bucket or pan. Minimize evaporation loss with snug fit lid and easily remove with comfortable recessed knobs. Keyed lid and base fit together to conveniently store lid under base. Use handy pour spout with melted ice, water, and solutions. Ergonomic handles safely transport bucket or pan from work area to sink. Deep internal cavity has enough room to add one of your racks. Tapered sides make stacking and storage compact.