Learn More
Epredia™ Cool-Cut with universal cassette clamp
Catalog No. 1407124 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Standard Clamp
Universal Cassette Clamp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-071-24 Universal Cassette Clamp
77-112-0 Standard Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-071-24 Supplier Epredia™ Supplier No. 771110
Add to Cart
Add to Cart
Maintain a cool, consistent temperature for paraffin blocks during the sectioning process with the Epredia™ Cool-Cut accessory.
  • Directly fits onto the specimen cylinder of the rotary microtome line
  • Ability to be equipped as a universal cassette clamp or standard specimen clamp
  • Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
  • Peltier device with integrated specimen clamp
  • Counter-balance weights
  • Power supply
Specifications
Product Type Universal Cassette Clamp
For Use With (Equipment) Epredia Microtomes
WARNING: Cancer - www.P65Warnings.ca.gov