Learn More
Med Vet International Cool Renewal™, Plastic Isolation Funnels, 12mm, J1401D7, 50/Pack
Catalog No. 5000581226
Encompass_Preferred
For use on fleshy tissue areas where a tight seal can be achieved. DO NOT use on head, face or neck. Disposable between patients. Plastic Isolation Funnels Disposable. 12mm funnel (qty 50)
Catalog No. 50-005-81226 Supplier Med Vet International Supplier No. JOR1401D7
For use on fleshy tissue areas where a tight seal can be achieved. DO NOT use on head, face or neck. Disposable between patients. Plastic Isolation Funnels Disposable. 12mm funnel (qty 50)