Learn More
Med Vet International Cool Renewal™, Plastic Isolation Funnels, 16mm, J1401D9, 50/Pack
Supplier: Med Vet International JOR1401D9
For use on fleshy tissue areas where a tight seal can be achieved. DO NOT use on head, face or neck. Disposable between patients. Plastic Isolation Funnels Disposable. 16mm funnel (qty 50)