Learn More
Lakeland Industries Cool Suit Coveralls
Supplier:  Lakeland Industries COL4284X
Encompass_Preferred
MicroMax NS Cool Suits are engineered with a breathable back panel to let hot air escape and keep the wearer cooler. COL428 coveralls are configured with a hood, elastic wrists, and ankles. Coming in a case of 25, sizes SM-5X, MicroMax NS Cool Suits are ideal for general maintenance, as well as protection against dirt, dust, grease, and grime.
Catalog No. 50-284-1204