Learn More
Lakeland Industries Cool Suit Coveralls
Catalog No. 502840801
Encompass_Preferred
MicroMax NS Cool Suits are engineered with a breathable back panel to let hot air escape and keep the wearer cooler. COL412 collared coveralls are configured with open wrists and ankles. Coming in a case of 25, sizes SM-5X, MicroMax NS Cool Suits are ideal for general maintenance, as well as protection against dirt, dust, grease, and grime.
Catalog No. 50-284-0801 Supplier Lakeland Industries Supplier No. COL412XL
MicroMax NS Cool Suits are engineered with a breathable back panel to let hot air escape and keep the wearer cooler. COL412 collared coveralls are configured with open wrists and ankles. Coming in a case of 25, sizes SM-5X, MicroMax NS Cool Suits are ideal for general maintenance, as well as protection against dirt, dust, grease, and grime.