Learn More
Heidolph™ Cool Trap Condenser for Laborota 20, Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900925 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-925 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-925 Supplier Heidolph™ Supplier No. 036251540
Add to Cart
Add to Cart
Request a Quote
Cool trap condenser allows distillate to re-condense. Condenser for low-boiling solvents; cooling by dry ice.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware set K and KC
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov