Learn More
Research Products International Corp CoolCaddy PCR Workstation, 1 each
SDP
Catalog No. NC1590388
Encompass
Keep samples cool for up to 5 hours on a benchtop. The CoolCADDY protects enzymes, reagents, and temperature sensitive samples when working with PCR samples. Ideal workstation for PCR protocols requiring multi dispensing of samples.  Offers highest level on enzyme protection and is essential for all protocols that require more than 30 minutes of benchtop prep work.
Simply prechill the CoolCADDY in freezer overnight and it's ready to use. CADDY holds 5 x 0.5 ml; 9 x 1.5-2.0 ml; 8 x 2.0 ml cryotubes; 1 x 96 well plate or 96 PCR tubes; and 1 x 50 ml reservoir.
Maintains 2 - 4°C for up to 5 hours
Keeps samples cool and not frozen
Holds vials, tubes, plates and reservoirs
Catalog No. NC1590388 Supplier Research Products International Corp Supplier No. 353120
Keep samples cool for up to 5 hours on a benchtop. The CoolCADDY protects enzymes, reagents, and temperature sensitive samples when working with PCR samples. Ideal workstation for PCR protocols requiring multi dispensing of samples.  Offers highest level on enzyme protection and is essential for all protocols that require more than 30 minutes of benchtop prep work.
Simply prechill the CoolCADDY in freezer overnight and it's ready to use. CADDY holds 5 x 0.5 ml; 9 x 1.5-2.0 ml; 8 x 2.0 ml cryotubes; 1 x 96 well plate or 96 PCR tubes; and 1 x 50 ml reservoir.
Maintains 2 - 4°C for up to 5 hours
Keeps samples cool and not frozen
Holds vials, tubes, plates and reservoirs