Learn More
ATCC CoolCell LX; Cryopreservation; Container
Catalog No. 502384003
Encompass
CoolCell LX Alcohol-free Cryopreservation container can be used in cryopreservation leading to optimal cell viability. CoolCell LX containers, in combination with a -80°C freezer or dry ice locker, will provide a freezing rate of -1°C per minute which is ideal for cryo-preservation of most cultured cell lines.
Catalog No. 50-238-4003 Supplier ATCC Supplier No. ACS6000
Add to Cart
Add to Cart
CoolCell LX Alcohol-free Cryopreservation container can be used in cryopreservation leading to optimal cell viability. CoolCell LX containers, in combination with a -80°C freezer or dry ice locker, will provide a freezing rate of -1°C per minute which is ideal for cryo-preservation of most cultured cell lines.