Learn More
LabScientific COOLCUBE™ MICROTUBE AND PCR PLATE COOLER Maintains 0°C for up to 4 hours, also for use as PCR workstation, two sides, ideal for: 1.5 & 2.0ml microtubes, 0.2ml PCR tubes & strips, 96 well PCR plates
Catalog No. NC1255071
Encompass
This product is specifically designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Simply place the unit in the freezer overnight and it’s ready for use, keeping samples cold, but not frozen. One side of the unit will hold 36×1.5/2.0ml tubes. Flip it over, and you have a PCR workstation. This side holds one 96 well PCR plate (or 96×0.2ml tubes) and ten 1.5/2.0ml tubes. Dimensions: 5.6 x 5.6 x 2.5 in
Catalog No. NC1255071 Supplier LabScientific Supplier No. R1000
This product is specifically designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Simply place the unit in the freezer overnight and it’s ready for use, keeping samples cold, but not frozen. One side of the unit will hold 36×1.5/2.0ml tubes. Flip it over, and you have a PCR workstation. This side holds one 96 well PCR plate (or 96×0.2ml tubes) and ten 1.5/2.0ml tubes. Dimensions: 5.6 x 5.6 x 2.5 in