Learn More
Thomas Scientific CoolCube Microtube and PCR Plate Cooler
SDP
Catalog No. NC1258278
Encompass_Preferred
The CoolCube is designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Constructed of a high strength polymer and a patented gel refrigerant, the CoolCube maintains temperature at approximately 0° C for up to four hours. Simply place the CoolCube in a freezer overnight and it’s ready for use, keeping samples cool and safe at the bench without messy ice buckets or wet tubes. One side of the CoolCube can be used as a PCR workstation with wells for 12 standard 1.5/2.0ml microtubes and one 96 well PCR plate or 0.2ml tubes/strips. The other side can be used to hold 36 standard 1.5/2.0ml microtubes.
Catalog No. NC1258278 Supplier Thomas Scientific Supplier No. 1227W01
The CoolCube is designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation. Constructed of a high strength polymer and a patented gel refrigerant, the CoolCube maintains temperature at approximately 0° C for up to four hours. Simply place the CoolCube in a freezer overnight and it’s ready for use, keeping samples cool and safe at the bench without messy ice buckets or wet tubes. One side of the CoolCube can be used as a PCR workstation with wells for 12 standard 1.5/2.0ml microtubes and one 96 well PCR plate or 0.2ml tubes/strips. The other side can be used to hold 36 standard 1.5/2.0ml microtubes.