Learn More
Cole-Parmer Masterflex Circuit for Solenoid Valves, 12 or 24 VDC input
Catalog No. NC0143183 Shop All Cole-Parmer Products
Encompass_Preferred
Preserve the life of your solenoid valves with this "hit and hold" circuit that steps down DC voltage to 1/3 of input voltage and minimizes heat generation.
Catalog No. NC0143183 Supplier Cole-Parmer Supplier No. 0135652
Preserve the life of your solenoid valves with this "hit and hold" circuit that steps down DC voltage to 1/3 of input voltage and minimizes heat generation.