Learn More
Electron Microscopy Sciences CoolCube™ CoolCube Microtube and PCR Plate Cooler
SDP
Catalog No. 501929985
Encompass_Preferred
The CoolCube is made to safely store samples on the lab bench, without degradation from temperature fluctuation. Made of a high strength polymer and a gel refrigerant, it will maintain a temperature of approximately 0 degree C for up to 4 hours.
Store the CoolCube overnight in the freezer and it will be ready to use. It will keep samples cool, but not frozen, without messy ice buckets or wet tubes.
With two sides, it can also be used as a PCR workstation. One side has wells for 10 standard 1.5/2.0 ml microtubes and one 96-well PCR plate or 0.2 ml tubes/strips. The other side can be used to hold 36 standard 1.5/2.0 ml microtubes.
Catalog No. 50-192-9985 Supplier Electron Microscopy Sciences Supplier No. 61957
The CoolCube is made to safely store samples on the lab bench, without degradation from temperature fluctuation. Made of a high strength polymer and a gel refrigerant, it will maintain a temperature of approximately 0 degree C for up to 4 hours.
Store the CoolCube overnight in the freezer and it will be ready to use. It will keep samples cool, but not frozen, without messy ice buckets or wet tubes.
With two sides, it can also be used as a PCR workstation. One side has wells for 10 standard 1.5/2.0 ml microtubes and one 96-well PCR plate or 0.2 ml tubes/strips. The other side can be used to hold 36 standard 1.5/2.0 ml microtubes.