Learn More
Sigma Aldrich Fine Chemicals Biosciences CoolCubeTM Microtube and P1EA
Catalog No. 502900500
Encompass
The CoolCube is designed to store samples safely on the lab bench without sample degradation due to temperatureincrease or fluctuation. Simply place the CoolCube in a freezer overnight and its ready for use keeping samples cold but not frozenmaintaining temperature at approximately 0 C for up to four hours.Maintains 0C for up to 4 hoursAlso for use as a PCR workstationTwo sides ideal for1.5 2.0ml microtubes0.2ml PCR tubes strips96 well PCR plates
Catalog No. 50-290-0500 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7425851EA
The CoolCube is designed to store samples safely on the lab bench without sample degradation due to temperatureincrease or fluctuation. Simply place the CoolCube in a freezer overnight and its ready for use keeping samples cold but not frozenmaintaining temperature at approximately 0 C for up to four hours.Maintains 0C for up to 4 hoursAlso for use as a PCR workstationTwo sides ideal for1.5 2.0ml microtubes0.2ml PCR tubes strips96 well PCR plates