Learn More
Ace Glass, Inc. Cooler, condensate, jacketed, poly-coated, model R220EX, glassware sets C, RB and R, part 41162
SDP
Catalog No. 50971491
Encompass
Borosilicate glass replacement poly-coated jacketed condensate cooler for use in rotary evaporator Model R220EX glassware sets C, RB & R; part 41162. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-491 Supplier Ace Glass, Inc. Supplier No. 396530
Borosilicate glass replacement poly-coated jacketed condensate cooler for use in rotary evaporator Model R220EX glassware sets C, RB & R; part 41162. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.