Learn More
Ace Glass, Inc. Cooler, condensate, jacketed, model R220EX, glassware sets R, RB and R, part 46510
SDP
Catalog No. 50971492
Encompass
Borosilicate glass replacement jacketed condensate cooler for use in rotary evaporator Model R220EX glassware sets C, RB & R; part 46510. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-492 Supplier Ace Glass, Inc. Supplier No. 396532
Borosilicate glass replacement jacketed condensate cooler for use in rotary evaporator Model R220EX glassware sets C, RB & R; part 46510. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.