Learn More
Ace Glass, Inc. Cooler, condensate, poly-coated, model R220, glassware sets C, RB and R
SDP
Catalog No. 50978237
Encompass
Borosilicate glass replacement poly-coated jacketed condensate cooler for use in rotary evaporator Model R220 glassware sets C, RB & R. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-978-237 Supplier Ace Glass, Inc. Supplier No. 396534
Borosilicate glass replacement poly-coated jacketed condensate cooler for use in rotary evaporator Model R220 glassware sets C, RB & R. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.