Learn More
Ace Glass, Inc. Cooler, condensate, model R220, glassware sets C, RB and R
SDP
Catalog No. 50971493
Encompass
Borosilicate glass replacement jacketed condensate cooler for use in rotary evaporator Model R220 glassware sets C, RB & R. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-493 Supplier Ace Glass, Inc. Supplier No. 396536
Borosilicate glass replacement jacketed condensate cooler for use in rotary evaporator Model R220 glassware sets C, RB & R. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.