Learn More
Polyscience IP-60 Immersion Probe Cooler, 1.5 in Rigid Coil Probe (60 to -20°C); 120V, 60Hz
SDP
Catalog No. NC1682307
Encompass_Preferred
PolyScience™ IP-60 Immersion Probe Cooler 1.5 Inch Diameter Rigid Coil Probe-60 to -20°C Working Temperature Range120V, 60HzFeatures:• Continuous cooling to -60°C• Designed to run at Maximum cooling• An economical alternative to dry ice or liquid nitrogen• Excellent for trapping applications, freeze drying, and rapidly cooling small volumes of liquids
Catalog No. NC1682307 Supplier Polyscience Supplier No. P60N2A101B
PolyScience™ IP-60 Immersion Probe Cooler 1.5 Inch Diameter Rigid Coil Probe-60 to -20°C Working Temperature Range120V, 60HzFeatures:• Continuous cooling to -60°C• Designed to run at Maximum cooling• An economical alternative to dry ice or liquid nitrogen• Excellent for trapping applications, freeze drying, and rapidly cooling small volumes of liquids