Learn More
Sklar Instruments Forceps, Cooley Vascular Tissue Forceps, Sklar 8 in., Blunt, Straight, 2X2 Rows of Nontraumatic Fine Teeth, Stainless Steel, EA.
SDP
Catalog No. 501109674 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Cooley Vascular Tissue Forceps, Sklar, Blunt, Straight, 2X2 Rows of Nontraumatic Fine Teeth, Stainless Steel, EA. Sklar Cooley Atraumatic Vascular Tissue Forceps are a thumb forceps used primarily in cardiothoracic procedures to grasp delicate tissue. The Cooley style 2x2 fine, atraumatic teeth cause little to no damage and the narrow tips can be placed in small, narrow areas. The thumb grip is slightly wider than the tips so the physician will have adequate control. Sklar operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar instruments meet the demands of our customers. This product is straight with 2x2 rows of fine, atraumatic teeth and is 8 inches long. Lifetime warranty against all manufacturer defects.
Catalog No. 50-110-9674 Supplier Sklar Instruments Supplier No. 526694
Forceps, Cooley Vascular Tissue Forceps, Sklar, Blunt, Straight, 2X2 Rows of Nontraumatic Fine Teeth, Stainless Steel, EA. Sklar Cooley Atraumatic Vascular Tissue Forceps are a thumb forceps used primarily in cardiothoracic procedures to grasp delicate tissue. The Cooley style 2x2 fine, atraumatic teeth cause little to no damage and the narrow tips can be placed in small, narrow areas. The thumb grip is slightly wider than the tips so the physician will have adequate control. Sklar operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar instruments meet the demands of our customers. This product is straight with 2x2 rows of fine, atraumatic teeth and is 8 inches long. Lifetime warranty against all manufacturer defects.