Learn More
Stoelting™ Coolight LED Illuminator
SDP
Catalog No. 10001047 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Single gooseneck
Catalog No. Model
10-001-047 Single gooseneck
1 options
Catalog No. 10-001-047 Supplier Stoelting™ Supplier No. 59253
Add to Cart
Add to Cart
Stoelting™ Coolight LED Illuminator featuers advantages over halogen and metal halide lamps including long life, energy efficiency, true light source, interchangeable fiber optic interface adapter, smaller package, minimal color temperature variation when dimmed and over lifetime, fast warm-up and output stabilization, and no high voltage power supply
Specifications
For Use With (Application) Neuroscience
Model Single gooseneck