Learn More
Bellco Glass, Inc. Cooling 2 Valve Assembly ,115V
SDP
Catalog No. 501946392
Encompass_Preferred
2 valve model, 115v. MADE IN THE USASupplied with:Plastic hose connector (1 per valve)Push fitting (1 per valve)M3 x 5 screw & #5 int. Lock-washer (2 per valve)(1) sst base plate(4) rubber feet(1) solenoid valve sub-assembly
Catalog No. 50-194-6392 Supplier Bellco Glass, Inc. Supplier No. 780801002
2 valve model, 115v. MADE IN THE USASupplied with:Plastic hose connector (1 per valve)Push fitting (1 per valve)M3 x 5 screw & #5 int. Lock-washer (2 per valve)(1) sst base plate(4) rubber feet(1) solenoid valve sub-assembly