Learn More
Bellco Glass, Inc. Cooling 3 Valve Assembly , 230V
SDP
Catalog No. 501946396
Encompass_Preferred
3 valve model, 230vSupplied with:Plastic hose connector (1 per valve)Push fitting (1 per valve)M3 x 5 screw & #5 int. Lock-washer (2 per valve)(1) sst base plate(4) rubber feet(1) solenoid valve sub-assembly. MADE IN THE USA
Catalog No. 50-194-6396 Supplier Bellco Glass, Inc. Supplier No. 780802003
3 valve model, 230vSupplied with:Plastic hose connector (1 per valve)Push fitting (1 per valve)M3 x 5 screw & #5 int. Lock-washer (2 per valve)(1) sst base plate(4) rubber feet(1) solenoid valve sub-assembly. MADE IN THE USA