Learn More
Grainger Cooling Bandana Blue One Size
Catalog No. 502767043
Encompass_Preferred
The Grainger Cooling Bandana is a blue polyester bandana that provides cooling relief when wet. It measures 21" x 21" and features an absorbent inner lining that retains water. To use, soak the bandana in cool water, wring out excess, and tie around neck for a cooling effect as the water evaporates. The bandana can be re-wet as needed for prolonged cooling. It is washable, reusable, and offers UPF 50+ sun protection. This versatile accessory can provide cooling relief while working, exercising, or enjoying outdoor activities.
Catalog No. 50-276-7043 Supplier Grainger Supplier No. 16V843
The Grainger Cooling Bandana is a blue polyester bandana that provides cooling relief when wet. It measures 21" x 21" and features an absorbent inner lining that retains water. To use, soak the bandana in cool water, wring out excess, and tie around neck for a cooling effect as the water evaporates. The bandana can be re-wet as needed for prolonged cooling. It is washable, reusable, and offers UPF 50+ sun protection. This versatile accessory can provide cooling relief while working, exercising, or enjoying outdoor activities.